Chemical resistant gloves, AlphaTec® 58-128
Supplier: Ansell
Certificates
About this item
These nylon gloves with thin nitrile coating provide protection against chemicals, oils and other irritant fluids, such as biocides, pesticides, fungicides, paints, inks and dyes.
- Flexible open weave knit liner and stretchy nitrile formulation coating for better dexterity, tactility and comfort
- 'Thin Nitrile Technology' formulation: Provides optimal flexibility and enables the hand to move naturally without constriction
- Ansell Grip Technology™ offers more precise handling of small oily or wet parts
- Extended cuff provides forearm splash protection
Ideal for agriculture applications, chemical handlings, light assembly in chemical environments, refining, metal working with corrosives, packaging and filling.
Certifications: EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A); Cat. III. Food contact approved.
Specifications
- Catalog No:
- 112-4392
- 112-4390
- 112-4391
- 112-4393
- 112-4389
- Model:
- 58-128
- 58-128
- 58-128
- 58-128
- 58-128
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Ansell Grip Technology™
- Ansell Grip Technology™
- Ansell Grip Technology™
- Ansell Grip Technology™
- Ansell Grip Technology™
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A)
- EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A)
- EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A)
- EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A)
- EN ISO 374-1:2016 Type A (JKOPST), EN ISO 374-5:2016, EN 388:2016 (X121A)