Chemical resistant gloves, AlphaTec® 38-612
Supplier: Ansell
Certificates
About this item
These gloves made from Viton/Butyl provide excellent resistance to the most aggressive chemicals, such as aliphatic, halogenated and aromatic hydrocarbons as well as concentrated mineral acids.
- Very flexible and provide excellent dexterity
- Natural, curved ergonomic shape and soft feel
- Easy donning and good grip
Ideal for use in chemical plants, chemical transportation and processing, oil refining and petrochemicals, disaster response and HazMat work.
Certifications: EN ISO 374-1:2016 Type A (BCFLPST), EN ISO 374-5:2016, EN 388:2016 (1010X); Cat. III
Specifications
- Catalog No:
- 112-4388
- 112-4386
- Model:
- 38-612
- 38-612
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Finish:
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1:2016 Type A (BCFLPST), EN ISO 374-5:2016, EN 388:2016 (1010X)
- EN ISO 374-1:2016 Type A (BCFLPST), EN ISO 374-5:2016, EN 388:2016 (1010X)