ESD safety ankle boots, lace-up, Trainers, Sander Pro GTX
Supplier: ELTEN
Certificates
About this item
100% metal- and leather-free, lightweight ESD safety ankle boots made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
- Gore-Tex® climate membrane
- Full length fleece inlay sole (ESD Pro), black
- TPU scuff cap protects the leather upper
- Metal-free puncture resistant midsole
- Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifications: EN 61340-5-1 (ESD); EN ISO 20345; S3.
Specifications
- Catalog No:
- 114-6259
- 114-6260
- 114-6258
- 114-6265
- 114-6256
- 114-6257
- 114-6261
- 114-6262
- 114-6263
- 114-6264
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001