Orders LinkContactUsLinkComponent
Bandage mull
Bandage mull
Supplier:   HARTMANN PAUL
Bandage mull
Supplier:  HARTMANN PAUL
Certificates
Bandage mull
2 Options Available Below
Product Details & Documents
Mull for making individual bandages and wound dressings.
  • 20 threads
  • Laid 8-fold in zigzag layers to a width of 10 cm
  • Good purity and absorbency
 : EN 14079
Product Family Options
Product Information
  • Availability
    Price
  • HART206406/3
    Gauze, 20-ply, zigzag layers, 80 cm wide, 10 m long
    Gauze, 20-ply, zigzag layers, 80 cm wide, 10 m long
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items - 48,20 €
    48,20 €
    1 * 8 items
       48,20 €
  • HART2064072
    Gauze, 20-ply, zigzag layers, 80 cm wide, 40 m long
    Gauze, 20-ply, zigzag layers, 80 cm wide, 40 m long
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 155,00 €
    155,00 €
    1 * 1 items
       155,00 €