ContactUsLinkComponent
Membrane disc filters
Catalog # HACH1353001
Supplier:  Hach
undefined
Membrane disc filters
Catalog # HACH1353001
Supplier:  Hach
Specifications
  • Ø:
    47 mm
  • Pore size:
    0,45 µm
  • Type:
    GN-6, grid, individually packed, without pad
  • Pk:
    200
Specifications
About this item
These GN-metrical membrane filters are ideal for testing recreational water, wastewater and drinking water. They meets U.S. Environmental Protection Agency (USEPA) and American Society for Testing and Materials (ASTM) specifications to ensure complete retention and maximum recovery of bacteria.
  • Convenient and pre-sterilised
  • Gridded design