Anti-Acetyl Lysine (AcK) Rabbit Polyclonal Antibody
Catalog # BSENR-1547-100
Supplier: Biosensis
Specifications
- Antibody type:Primary
- Antigen name:Acetyl-Lysine
- Antigen symbol:AcK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Immunogen:Acetylated Lysine OVA
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Lysine acetylation of histones and non-histone proteins plays an important part in many cellular processes such as chromatin and nuclear signaling, transcription, gene silencing, cell cycle progression, apoptosis, differentiation, DNA replication and repair.
Binds to proteins with acetylated lysine residues.
Application Information:
WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Acetyl Lysine (AcK)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: