Order Entry
Export
ContactUsLinkComponent
Anti-Ubiquitin C Terminal Hydrolase 1 Mouse Monoclonal Antibody [clone: BH7]
Anti-Ubiquitin C Terminal Hydrolase 1 Mouse Monoclonal Antibody [clone: BH7]
Catalog # BSENM-1407-100
Supplier:  Biosensis
undefined
Anti-Ubiquitin C Terminal Hydrolase 1 Mouse Monoclonal Antibody [clone: BH7]
Catalog # BSENM-1407-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin C Terminal Hydrolase 1
  • Clonality:
    Monoclonal
  • Clone:
    BH7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    Ubiquitin carboxyl-terminal hydrolase isozyme L1
  • Immunogen:
    Recombinant full length human Ubiquitin C Terminal Hydrolase 1 (UCHL1) purified from E. coli.
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. The enzyme also binds to free monoubiquitin and may prevent its degradation in lysosomes (ref: SWISSPROT).

The specificity of this antibody has been confirmed by WB. This antibody detects ~24 kDa UCHL1 enzyme. Suitable control tissue is rat spinal cord, brain, SHSY-5Y or HEK293 cell extract.
Application Information:
Western Blotting (WB), Immunocytochemistry (IC) and Immunohistochemistry (IH). A dilution of 1:20,000 is recommended for WB. A dilution of 1:500 - 1:2,000 is recommended for IC and IH. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ubiquitin C Terminal Hydrolase 1
Clonality: Monoclonal
Clone: BH7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Bovine