- Antibody type:Primary
- Antigen name:Synuclein alpha
- Antigen symbol:SNCA
- Clonality:Monoclonal
- Clone:3H9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rodent,Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:PARK1|PARK4|NACP|PD1
- Immunogen:Recombinant full length human alpha synuclein expressed and purified from E. coli
- Size:100 µl
- Pk:100 µl
Alpha synuclein is an abundant 140 amino acid neuronal protein, expressed primarily at presynaptic terminals in the central nervous system. FUNCTION: May be involved in the regulation of dopamine release and transport. Soluble protein, normally localized primarily at the presynaptic region of axons, which can form filamentous aggregates that are the major non amyloid component of intracellular inclusions in several neurodegenerative diseases (synucleinopathies). Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase 3 activation. TISSUE SPECIFICITY: Expressed principally in brain but is also expressed in low concentrations in all tissues examined except in liver. Concentrated in presynaptic nerve terminals.SUBUNIT: Soluble monomer which can form filamentous aggregates. Interacts with UCHL1. Interacts with phospholipase D and histones. SUBCELLULAR LOCATION: Cytoplasm. Membrane. Nucleus. Note=Membrane-bound in dopaminergic neurons. Also found in the nucleus. ALTERNATIVE PRODUCTS: 3 named isoforms produced by alternative splicing. Additional isoforms seem to exist.
The specificity of this antibody has been confirmed by WB. This antibody detects ~15 kDa alpha synuclein protein.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:1,000 - 1:5,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: alpha Synuclein
Clonality: Monoclonal
Clone: 3H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rodent