Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin C Terminal Hydrolase 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgY
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:Multiple points of reactivity likely because of the polyclonal nature of the antibody
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Ubiquitin carboxyl-terminal hydrolase isozyme L1
- Immunogen:Recombinant full length human Ubiquitin C Terminal Hydrolase 1 (UCHL1) purified from E. coli.
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Ubiquitin C-terminal hydrolase 1 (UCHL1) has several other names, such as ubiquitin carboxyl esterase L1, ubiquitin thiolesterase, neuron-specific protein PGP9.5 and Park5. It was originally identified as a major component of the neuronal cytoplasm from 2-dimensional gel analysis of brain tissues, and was given the name PGP9.5 (1). The protein is extremely abundant, and was estimated to be present at a concentration of 200-500 µg/g wet weight, representing a major protein component of neuronal cytoplasm (1). This has been claimed to represent 1-2% of total brain protein. It was later found that a ubiquitin C-terminal hydrolase enzyme activity was associated with the PGP9.5 protein, resulting in the renaming of PGP9.5 to ubiquitin C-terminal hydrolase 1.This enzyme is a thiol protease that recognizes and hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. The enzyme also binds to free monoubiquitin and may prevent its degradation in lysosomes.
The specificity of this antibody has been confirmed by WB. This antibody detects ~24 kDa UCHL1 enzyme. Suitable control tissue is rat spinal cord or peripheral nerve homogenate. In cell staining, the antibody stains neurons strongly and specifically, and the staining is concentrated in the cell bodies, though some does extend into the dendrites.
Application Information:
Western Blotting (WB) and Immunocytochemistry (IC). A dilution of 1:5,000 - 1:10,000 is recommended for WB. A dilution of 1:500-1,000 is recommended for IC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Ubiquitin C Terminal Hydrolase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat, Mouse, Bovine, Pig