Anti-mouse adrenergic b3 receptor Chicken Polyclonal Antibody
Catalog # BSENC-1529-100
Supplier: Biosensis
Specifications
- Antibody type:Primary
- Antigen name:Adrenergic b3 Receptor
- Antigen symbol:ADRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:Mouse beta 3 Adrenergic Receptor (386-400 aa).
- Cross adsorption:No
- Format:Liquid
- Immunogen:A synthetic peptide from mouse beta 3 Adrenergic Receptor (386-400 aa).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Beta-adrenergic receptors are multi-pass membrane proteins that belong to the G-protein coupled receptor 1 family. Their function is to mediate the catecholamine-induced activation of adenylate cyclase through the action of G-proteins (ref: SWISSPROT).
This antibody detects ADRB3 in mouse and rat at approx 44 kDa.
Application Information:
WB and ELISA. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: mouse adrenergic b3 receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse beta 3 Adrenergic Receptor (386-400 aa).
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat