Order Entry
Export
ContactUsLinkComponent
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Catalog # BSENM-1314-100
Supplier:  Biosensis
undefined
Anti-Myc Tag Mouse Monoclonal Antibody [clone: BMYCA7]
Catalog # BSENM-1314-100
Supplier:  Biosensis

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    MYC Tag
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Clone:
    BMYCA7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Epitope:
    EQKLISEEDL
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A synthetic peptide (EQKLISEEDL) coupled to KLH.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The Myc tag contains the amino acids Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu (E-Q-K-L-I-S-E-E-D-L) corresponding to amino acids 410-419 of human Myc. This tag is widely used for monitoring expression of recombinant proteins in bacteria, insect and mammalian cells.

Antibody detects over-expressed proteins containing Myc epitope tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunoprecipitation (IP). Suggested starting dilutions as follows: WB at 1:1000, IP at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: Myc Tag
Clonality: Monoclonal
Clone: BMYCA7
Conjugation: Unconjugated
Epitope: EQKLISEEDL
Host: Mouse
Isotype:
Reactivity: