Specifications
- Antibody type:Primary
- Antigen name:Arg3.1
- Antigen symbol:ARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:C terminal of Human Arg3.1 protein
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:A synthetic peptide from the C terminal of human Arg3.1 protein (ARC protein) conjugated to Blue Carrier Protein. The sequence is homologous with mouse and rat form of Arg3.1.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Arc (also termed activity-regulated cytoskeleton-associated protein or Arg3.1), is an effector immediate early gene whose upregulation has been demonstrated during events of synaptic plasticity. Arg3.1 expression is detectable in neuronal cell bodies and dendrites in the brain regions including striatum and cortex hippocampus, hypothalamus, amygdala.
IHC. A concentration of of 2-4 µg/ml is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Other applications and other species have not been tested.
Application Information:
IHC. A concentration of of 2-4 µg/ml is recommended for this application. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Arg3.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C terminal of Human Arg3.1 protein
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse