Order Entry
Export
ContactUsLinkComponent
Anti-cFOS Sheep Polyclonal Antibody
Anti-cFOS Sheep Polyclonal Antibody
Catalog # BSENS-045-50
Supplier:  Biosensis
undefined
Anti-cFOS Sheep Polyclonal Antibody
Catalog # BSENS-045-50
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proto-oncogene protein cFOS
  • Antigen symbol:
    cFOS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Hamster,
    Rabbit,
    Rat
  • Epitope:
    aa 2-17 c-Fos
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Immunogen:
    A synthetic peptide (MFSGFNADYEASSSRC; aa 2-17) conjugated to diphtheria toxoid has been used as the immunogen. The peptide is homologous with the corresponding sequence derived from cFos protein human, rat, mouse, hamster and cat.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

FUNCTION: Nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. Has a critical function in regulating the development of cells destined to form and maintain the skeleton. It is thought to have an important role in signal transduction, cell proliferation and differentiation. SUBUNIT: Heterodimer. Interacts with DSIPI; this interaction inhibits the binding of active AP1 to its target DNA. Interacts with MAFB. SUBCELLULAR LOCATION: Nucleus. INDUCTION: C-fos expression increases upon a variety of stimuli, including growth factors, cytokines, neurotransmitters, polypeptide hormones, stress and cell injury. SIMILARITY: Belongs to the bZIP family. Fos subfamily. SIMILARITY: Contains 1 bZIP domain

This antiserum shows a high level of specificity for cFOS confirmed by immunohostochemistry.
Application Information:
IHC, WB. A concentration of 1 to 5 µg/ml is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

IH on PFA fixed, frozen tissues. Not yet tested on formalin fixed, paraffin embedded tissue but expected to react. Affinity purified antibody is extremely powerful. A concentration of 1 -5 µg/ml is recommended most uses with short (1-8 hour) incubations. If using enhanced brightfield or amplified detection methods dilutions and long primary antibody incubations dilutions will need to be increased substantially to inhibit background staining. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Not recommended for western blotting applications. Mouse monoclonal antibody BSENM-1752-100 or rabbit polyclonal antibody BSENR-1751-50 are excellent alternatives for western blotting.

Type: Primary
Antigen: cFOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa 2-17 c-Fos
Host: Sheep
Isotype: IgG
Reactivity: Rat, Rabbit, Hamster