Anti-CCL5 Mouse Polyclonal Antibody
Catalog # ABNOH00006352-B02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 5
- Antigen symbol:CCL5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6352
- Antigen synonyms:RANTES|SCYA5|SIS-delta|eoCP|TCP228|D17S136E|SISd
- Amino acid number:1 to 91
- Storage buffer:No additive
- Sequence:MKVSAAALAVILIATALCAPASASPYSSDTTPCCFAYIARPLPRAHIKEYFYTSGKCSNPAVVFVTRKNRQVCANPEKKWVREYINSLEMS
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CCL5 (AAH08600, 1 a.a. ~ 91 a.a) full-length human protein.
- Pk:50 µl
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human CCL5 protein.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: CCL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: