Anti-MIPOL1 Rabbit Polyclonal Antibody
ABNOPAB28729
: Abnova
- Antibody type:Primary
- Antigen name:Mirror-image Polydactyly 1
- Antigen symbol:MIPOL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:145282
- Storage buffer:PBS, pH 7,2 (40% glycerol, 0,02% sodium azide).
- Sequence:KTAALVEEVYFAQKERDEAVMSRLQLAIEERDEAIARAKHMEMSLKVLENINPEENDMTLQELLNRINNADTGIAIQKNGAIIVDRIYKTKECKMRITAEEMSALIEERDAALSKCKRLEQELHHVKEQNQTSANNMRHLTAENNQ
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human MIPOL1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against recombinant MIPOL1.
Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:100-1:250
Type: Primary
Antigen: MIPOL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: