Anti-RPL11 Rabbit Polyclonal Antibody
Catalog # ABNOPAB28685
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein L11
- Antigen symbol:RPL11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Cross adsorption:No
- Format:Liquid
- Gene ID:6135
- Antigen synonyms:CLL-associated antigen KW-12|RP11-223J15.3|GIG34|DBA7|L11|cell growth-inhibiting protein 34|60S ribosomal protein L11
- Storage buffer:PBS, pH 7,2 (40% glycerol, 0,02% sodium azide).
- Sequence:RAAKVLEQLTGQTPVFSKARYTVRSFGIRRNEKIAVHCTVRGAKAEEILEKGLKVREYELRKNNFSDTGNFGFGIQEHIDLGIKYDPSIGIYGLDFYVVLGRPGFSIADKKRRTGCIGAKHRISKEE
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human RPL11.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant RPL11.
Recommended Dilutions: Immunohistochemistry: 1:20-1:50
Type: Primary
Antigen: RPL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: