Anti-KIF5B Mouse Monoclonal Antibody [clone: 2A11]
ABNOH00003799-M01
: Abnova
- Antibody type:Primary
- Antigen name:kinesin family member 5B
- Antigen symbol:KIF5B
- Clonality:Monoclonal
- Clone:2A11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3799
- Antigen synonyms:UKHC|KNS|KINH|KNS1|HEL-S-61
- Amino acid number:864 to 962
- Storage buffer:1x PBS, pH 7,4
- Sequence:EKRLRATAERVKALESALKEAKENASRDRKRYQQEVDRIKEAVRSKNMARRGHSAQIAKPIRPGQHPAASPTHPSAIRGGGAFVQNSQPVAVRGGGGKQ
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:KIF5B (NP_004512.1, 864 a.a. ~ 962 a.a) partial recombinant protein with GST tag.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against a partial recombinant KIF5B.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: KIF5B
Clonality: Monoclonal
Clone: 2A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: