Anti-CGA Mouse Monoclonal Antibody [clone: HCGa/53]
Catalog # ABNOMAB12127
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein hormones, alpha polypeptide
- Antigen symbol:CGA
- Clonality:Monoclonal
- Clone:HCGa/53
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1081
- Antigen synonyms:CG-ALPHA|FSHA|LHA|GPHa|HCG|TSHA|GPHA1
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide).
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CGA.
- Purification:Protein A/G purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant human CGA.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CGA
Clonality: Monoclonal
Clone: HCGa/53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: