Anti-SLC12A1 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ABNOPAB29052
Supplier: Abnova
Specifications
- Antigen name:Solute Carrier Family 12 Member A1
- Antigen symbol:Solute carrier family 12, member 1
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- Host:Rabbit
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:25065
- Antigen synonyms:Nkcc2|BSC1|NKCC2
- Amino acid number:33 to 55
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 33-55 at N-terminus of rat Slc12a1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.
Recommended Dilutions: Western Blot: 1:1000
Type:
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: