Anti-TXNDC5 Mouse Monoclonal Antibody (ATTO 488) [clone: 2E7/7]
ABNOMAB11784
: Abnova
- Antibody type:Primary
- Antigen name:Thioredoxin domain containing 5
- Antigen symbol:TXNDC5
- Clonality:Monoclonal
- Clone:2E7/7
- Conjugation:ATTO 488
- Host:Mouse
- Isotype:IgG2a
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:81567
- Antigen synonyms:PDIA15|UNQ364|HCC2|STRF8|ERP46|HCC-2|ENDOPDI
- Amino acid number:52 to 63
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Sequence:ADGEDGQDPHSK
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids of 52-63 of human TXNDC5.
- Purification:Protein G Purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.
Recommended Dilutions: Western Blot: 1:200
Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2E7/7
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: