Anti-GRIN1 Mouse Monoclonal Antibody (ATTO 390) [clone: S308-48]
ABNOMAB11711
: Abnova
- Antibody type:Primary
- Antigen name:Glutamate [NMDA] receptor subunit zeta-1
- Antigen symbol:GRIN1
- Clonality:Monoclonal
- Clone:S308-48
- Conjugation:ATTO 390
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:24408
- Antigen synonyms:NMDAR1|NMD-R1|MRD8|GluN1|NR1|NMDA1
- Amino acid number:42 to 361
- Storage buffer:PBS pH 7,4 (50% glycerol and 0,09% sodium azide).
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protein corresponding to amino acids 42-361 at extracellular N-terminus of rat Grin1.
- Purification:Protein G purification
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: GRIN1
Clonality: Monoclonal
Clone: S308-48
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: