Anti-CD5 Mouse Monoclonal Antibody [clone: CD5-54-F6]
Catalog # ABNOMAB11490
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD5 molecule
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:CD5-54-F6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:liquid
- Gene ID:921
- Antigen synonyms:CD5|LEU1|Lymphocyte antigen T1/Leu-1|T-cell surface glycoprotein CD5|T1
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to the intracellular region of human CD5.
- Purification:Protein A/G affinity chromatography
- Size:100 µg
- Pk:100 µg
Specifications
About this item
mouse monoclonal antibody raised against synthetic peptide of CD5.
Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunohistochemistry: 0.5-1 ?g/ml for 30 min at RT; Immunoprecipitation: 0.5-1 ?g/500 ?g protein; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: CD5-54-F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: