Anti-POMC Mouse Monoclonal Antibody [clone: AH26 + 57]
ABNOMAB11360
: Abnova
- Antibody type:Primary
- Antigen name:proopiomelanocortin
- Antigen symbol:POMC
- Clonality:Monoclonal
- Clone:AH26 + 57
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:5443
- Antigen synonyms:MSH|NPP|POC|CLIP|LPH|ACTH
- Amino acid number:1 to 24
- Storage buffer:PBS, pH 7,4 (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-24 of human POMC.
- Purification:Protein A/G affinity chromatography
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of POMC.
Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunofluorescence: 0.5-1 ?g/ml; Immunohistochemistry: 0.5-1 ?g/ml for 30 min at RT; Immunoprecipitation: 0.5-1 ?g/500 ?g protein lysate; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: POMC
Clonality: Monoclonal
Clone: AH26 + 57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa + IgG1 kappa
Reactivity: Human