Anti-CCR7 Rat Monoclonal Antibody [clone: 4B12]
ABNOMAB10929
: Abnova
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 7
- Antigen symbol:CCR7
- Clonality:Monoclonal
- Clone:4B12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:12775
- Antigen synonyms:Epstein-Barr Virus-Induced G-Protein Coupled Receptor 1|MIP-3 Beta Receptor |EBI1|CCR-7|Chemokine (C-C Motif) Receptor 7|CD197|CMKBR7
- Storage buffer:aqueous buffer (0,09% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Mouse Ccr7.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µg
Rat monoclonal antibody raised against Ccr7.
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse