Anti-FAM46C Mouse Polyclonal Antibody
Catalog # ABNOH00054855-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Family with sequence similarity 46, member C
- Antigen symbol:FAM46C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:54855
- Amino acid number:1 to 391
- Storage buffer:1x PBS, pH 7,4
- Sequence:MAEESSCTRDCMSFSVLNWDQVSRLHEVLTEVVPIHGRGNFPTLEITLKDIVQTVRSRLEEAGIKVHDVRLNGSAAGHVLVKDNGLGCKDLDLIFHVALPTEAEFQLVRDVVLCSLLNFLPEGVNKLKISPVTLKEAYVQKLVKVCTDTDRWSLISLSNKNGKNVELKFVDSIRRQFEFSVDSFQIILDSLLFFYDCSNNPISEHFHPTVIGESMYGDFEEAFDHLQNRLIATKNPEEIRGGGLLKYSNLLVRDFRPTDQEEIKTLERYMCSRFFIDFPDILEQQRKLETYLQNHFAEEERSKYDYLMILRRVVNESTVCLMGHERRQTLNLISLLALRVLAEQNIIPSATNVTCYYQPAPYVSDGNFSNYYVAHPPVTYSQPYPTWLPCN
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FAM46C (NP_060179.2, 1 a.a. ~ 391 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human FAM46C protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FAM46C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human