Anti-CYP51A1 Rabbit Polyclonal Antibody
Catalog # ABNOH00001595-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 51, Subfamily A, Polypeptide 1
- Antigen symbol:CYP51A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1595
- Antigen synonyms:CYP51|CYPL1|P450L1|LDM|CP51|P450-14DM
- Amino acid number:1 to 509
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CYP51A1 (ABM85421.1, 1 a.a. ~ 509 a.a) full-length human protein.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against a full-length human CYP51A1 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CYP51A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human