Order Entry
Export
ContactUsLinkComponent
Anti-CK8 Mouse Monoclonal Antibody [clone: C-43]
Anti-CK8 Mouse Monoclonal Antibody [clone: C-43]
Catalog # ABNOMAB0856
Supplier:  Abnova
Anti-CK8 Mouse Monoclonal Antibody [clone: C-43]
Catalog # ABNOMAB0856
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 8
  • Antigen symbol:
    CK8
  • Clonality:
    Monoclonal
  • Clone:
    C-43
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Rabbit,
    Sheep,
    Human,
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3856
  • Antigen synonyms:
    cytokeratin 8|Cytokeratin endo A|CYK8|Keratin|KRT8|type II cytoskeletal 8|K8|KRT2-8|CK 8|K2C8|CK8
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified KRT8 from cytoskeleton of human HeLa cell line.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native KRT8.

Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: C-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig, Rabbit, Sheep