Anti-CK7 Mouse Monoclonal Antibody [clone: C-46]
Catalog # ABNOMAB0859
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 7
- Antigen symbol:CK7
- Clonality:Monoclonal
- Clone:C-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3855|3872
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified KRT7/KRT17 from cytoskeleton of human HeLa cell line.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native KRT7/KRT17.
Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: C-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig