Anti-TNFRSF10A Mouse Monoclonal Antibody [clone: 32A1380]
ABNOMAB0054
: Abnova
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily, Member 10A
- Antigen symbol:TNFRSF10A
- Clonality:Monoclonal
- Clone:32A1380
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8797
- Antigen synonyms:TRAILR1|DR4|APO2|CD261|TRAILR-1
- Amino acid number:1 to 20
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-20 of human TNFRSF10A.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of TNFRSF10A.
Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: 32A1380
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human