Anti-CGB Mouse Monoclonal Antibody [clone: HCG-61]
Catalog # ABNOMAB0865
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein hormones, beta polypeptide
- Antigen symbol:CGB
- Clonality:Monoclonal
- Clone:HCG-61
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:1082
- Antigen synonyms:CGB3|CG-beta|chorionic gonadotropin beta 3 subunit|chorionic gonadotropin beta subunit|chorionic gonadotropin beta chain|CGB8|choriogonadotropin subunit beta|CGB5|chorionic gonadotrophin chain beta|hCGB|CGB7
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human CGB.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CGB.
Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: HCG-61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human