Anti-TUP1 Mouse Monoclonal Antibody [clone: 10]
Catalog # ABNOMAB1790
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:TUP1
- Clonality:Monoclonal
- Clone:10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Yeast
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:850445
- Antigen synonyms:SFL2|AMM1|CRT4|CYC9|AAR1|YCR084C|ROX4|AER2|UMR7|FLK1
- Amino acid number:1 to 200
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-200 of Yeast TUP1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant TUP1.
Type: Primary
Antigen: TUP1
Clonality: Monoclonal
Clone: 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Yeast