Order Entry
Export
ContactUsLinkComponent
Anti-BrdU Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MoBu1]
Anti-BrdU Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MoBu1]
Catalog # ABNOMAB1828
Supplier:  Abnova
Anti-BrdU Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MoBu1]
Catalog # ABNOMAB1828
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bromodeoxyuridine
  • Antigen symbol:
    BrdU
  • Clonality:
    Monoclonal
  • Clone:
    MoBu1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    BrdU conjugated with Haemocyanin.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against BrdU.

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: MoBu1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype: IgG1
Reactivity: