Order Entry
Export
ContactUsLinkComponent
 
Anti-CCNE1 Mouse Monoclonal Antibody [clone: 168]
  ABNOMAB1856
 :  Abnova
Anti-CCNE1 Mouse Monoclonal Antibody [clone: 168]
  ABNOMAB1856
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin E1
  • Antigen symbol:
    CCNE1
  • Clonality:
    Monoclonal
  • Clone:
    168
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Monkey,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    898
  • Antigen synonyms:
    G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length human CCNE1.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against full length recombinant CCNE1.

Type: Primary
Antigen: CCNE1
Clonality: Monoclonal
Clone: 168
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey