Order Entry
Export
ContactUsLinkComponent
 
Anti-HSPB1 Mouse Monoclonal Antibody [clone: G3.1]
  ABNOMAB1889
 :  Abnova
Anti-HSPB1 Mouse Monoclonal Antibody [clone: G3.1]
  ABNOMAB1889
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 27kDa protein 1
  • Antigen symbol:
    HSPB1
  • Clonality:
    Monoclonal
  • Clone:
    G3.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3315
  • Antigen synonyms:
    SRP27|HSP27|HSP28|Hsp25|HEL-S-102|HS.76067|HMN2B|CMT2F
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified human HSPB1.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against native HSPB1.

Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: G3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human