Anti-IL7 Mouse Monoclonal Antibody [clone: 17-16F]
Catalog # ABNOMAB1311
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 7
- Antigen symbol:IL7
- Clonality:Monoclonal
- Clone:17-16F
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:3574
- Antigen synonyms:IL-7|IL7|Interluekin-7|Interluekin 7|IL 7|interluekin
- Storage buffer:PBS
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human IL7.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant IL7.
Recommended Dilutions: ELISA: 1:5000
Type: Primary
Antigen: IL7
Clonality: Monoclonal
Clone: 17-16F
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse