Anti-H5 NS Mouse Monoclonal Antibody [clone: 41]
Catalog # ABNOMAB10121
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Influenza A H5 NS Protein
- Antigen symbol:H5 NS
- Clonality:Monoclonal
- Clone:41
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified non-structural Influenza A H5 protein.
- Purification:Protein G purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native Influenza A H5 NS protein.
Type: Primary
Antigen: H5 NS
Clonality: Monoclonal
Clone: 41
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: