Anti-VCL Mouse Monoclonal Antibody [clone: VIN-54]
ABNOMAB1490
: Abnova
- Antibody type:Primary
- Antigen name:Vinculin
- Antigen symbol:VCL
- Clonality:Monoclonal
- Clone:VIN-54
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:7414
- Antigen synonyms:MVCL|Vinculin|CMH15|HEL114|Vinculin |MV|Metavinculin|CMD1W
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human uterus VCL.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native VCL.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: Frozen section) (2-4 ?g/ml
Type: Primary
Antigen: VCL
Clonality: Monoclonal
Clone: VIN-54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat