Order Entry
Export
ContactUsLinkComponent
Anti-CH Mouse Monoclonal Antibody [clone: CP-19]
Anti-CH Mouse Monoclonal Antibody [clone: CP-19]
Catalog # ABNOMAB1512
Supplier:  Abnova
Anti-CH Mouse Monoclonal Antibody [clone: CP-19]
Catalog # ABNOMAB1512
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calponin
  • Antigen symbol:
    CNN
  • Clonality:
    Monoclonal
  • Clone:
    CP-19
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Turkey,
    Chicken,
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    CNN3|Calponin 1 basic smooth muscle|CNN2|CNN1
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified turkey Calponin.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against native calponin.

Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.5-1 ?g/ml

Type: Primary
Antigen: CH
Clonality: Monoclonal
Clone: CP-19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Rat, Turkey