Anti-MYHSA1 Mouse Monoclonal Antibody [clone: MY-38]
Catalog # ABNOMAB1522
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Myosin
- Antigen symbol:MYHSA1
- Clonality:Monoclonal
- Clone:MY-38
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:MGC133384|MYHa|MYHSA1|MyHC-2X/D
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified rabbit myosin.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native myosin.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1.5-2 ?g/ml
Type: Primary
Antigen: MYHSA1
Clonality: Monoclonal
Clone: MY-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat