Anti-TPM1 Mouse Monoclonal Antibody [clone: TM-33]
Catalog # ABNOMAB1530
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tropomyosin 1
- Antigen symbol:TPM1
- Clonality:Monoclonal
- Clone:TM-33
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:alpha tropomyosin|TMSA|hypertrophic 3|alpha-tropomyosin|sarcomeric tropomyosin kappa|C15orf13|tropomyosin alpha-1 chain|tropomyosin-1|cardiomyopathy|CMD1Y|CMH3
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified chicken gizzard tropomyosin.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native tropomyosin.
Recommended Dilutions: Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: TPM1
Clonality: Monoclonal
Clone: TM-33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Mouse, Rat