Order Entry
Export
ContactUsLinkComponent
Anti-TPM1 Mouse Monoclonal Antibody [clone: TM-36]
Anti-TPM1 Mouse Monoclonal Antibody [clone: TM-36]
Catalog # ABNOMAB1531
Supplier:  Abnova
Anti-TPM1 Mouse Monoclonal Antibody [clone: TM-36]
Catalog # ABNOMAB1531
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tropomyosin 1
  • Antigen symbol:
    TPM1
  • Clonality:
    Monoclonal
  • Clone:
    TM-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Chicken
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    alpha tropomyosin|TMSA|hypertrophic 3|alpha-tropomyosin|sarcomeric tropomyosin kappa|C15orf13|tropomyosin alpha-1 chain|tropomyosin-1|cardiomyopathy|CMD1Y|CMH3
  • Storage buffer:
    1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
  • Storage temperature:
    Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified chicken gizzard tropomyosin.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against native tropomyosin.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry: Frozen section) (1-2 ?g/ml

Type: Primary
Antigen: TPM1
Clonality: Monoclonal
Clone: TM-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken