Anti-KIR2DL4 Mouse Monoclonal Antibody [clone: mAb#33]
Catalog # ABNOMAB10178
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 4
- Antigen symbol:KIR2DL4
- Clonality:Monoclonal
- Clone:mAb#33
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3805
- Antigen synonyms:KIR103|killer cell immunoglobulin-like receptor 2DL4|natural killer cell inhibitory receptor|killer Ig receptor|killer cell inhibitory receptor 103AS|MHC class I NK cell receptor KIR103AS|CD158 antigen-like family member D|NK cell receptor|KIR103AS|CD158D|G9P|KIR-103AS
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze. Avoid repeated freezing and thawing.
- Immunogen:NK3.3 cells and human KIR2DL4-Ig fusion protein.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against KIR2DL4.
Recommended Dilutions: Flow Cytometry: 1 ?g/ml
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb#33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human