Anti-BCL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Bcl2/100]
Catalog # ABNOMAB10228
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:B-cell lymphoma 2
- Antigen symbol:BCL2
- Clonality:Monoclonal
- Clone:Bcl2/100
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:596
- Antigen synonyms:PPP1R50|Bcl-2
- Amino acid number:41 to 54
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 41-54 of human BCL2.
- Purification:Size-exclusion chromatography purification
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of BCL2.
Recommended Dilutions: Flow Cytometry: 20 ?l reagent/100 ?l of whole blood or 10^6 cells
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: Bcl2/100
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human