Anti-CEL Monoclonal Antibody [clone: KNH-30]
Catalog # ABNOMAB1737
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:N(epsilon)-(carboxyethyl)lysine
- Antigen symbol:CEL
- Clonality:Monoclonal
- Clone:KNH-30
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:FAP|BSDL|FAPP|MODY8|LIPA|BAL|BSSL|CEase|CELL
- Storage buffer:PBS (0,1% proclin, 2.0% Block Ace)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CEL conjugated with BSA.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against CEL.
Recommended Dilutions: Immunohistochemistry: 5-10 ?g/ml; ELISA: 0.1-1.0 ?g/ml
Type: Primary
Antigen: CEL
Clonality: Monoclonal
Clone: KNH-30
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Mouse, Rat