Anti-DUSP13 Rabbit Polyclonal Antibody
ABNOPAB0294
: Abnova
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 13
- Antigen symbol:DUSP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Monkey,Chimpanzee,Human,Bovine,Dog
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:51207
- Antigen synonyms:MDSP|DUSP13A|BEDP|SKRP4|DUSP13B|TMDP
- Amino acid number:150 to 200
- Storage buffer:PBS (0,2% gelatin, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 150-200 of human DUSP13.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against partial recombinant DUSP13.
Type: Primary
Antigen: DUSP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Human, Monkey