Anti-CCNE1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB0820
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cyclin E1
- Antigen symbol:CCNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25729
- Antigen synonyms:G1/S-specific cyclin-E1|cyclin Et|cyclin Es|CCNE|CYCLE,CCNEcyclin Es
- Storage buffer:PBS (0,2% gelatin, 0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of rat Ccne1.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Rabbit polyclonal antibody raised against a synthetic peptide of Ccne1.
Recommended Dilutions: Western Blot: 1:200
Type: Primary
Antigen: CCNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat