Anti-BSA Sheep Polyclonal Antibody
200 μg
Catalog # ABNOPAB0903
Supplier: Abnova
Jump to:
Product Details & Documents
Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.
Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Documents
TDS_ABNOPAB0903.pdf
Datasheet
Specifications
- Size:200 μg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Host:Sheep
- Cross adsorption:No
- Antigen name:Bovine Serum Albumin
- Cat. No.:ABNOPAB0903
- Format:Liquid
- Immunogen:Bovine Serum Albumin, BSA.
- Conjugation:Unconjugated
- Antigen symbol:BSA
- Storage buffer:PBS (0,2% gelatin, 0,1% sodium azide)



