Anti-IBTK Rabbit Polyclonal Antibody
ABNOPAB10030
: Abnova
- Antibody type:Primary
- Antigen name:Inhibitor of Bruton agammaglobulinemia tyrosine kinase
- Antigen symbol:IBTK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25998
- Antigen synonyms:BTBD26|BTKI
- Amino acid number:1205 to 1217
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1205-1217 of human IBTK.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of IBTK.
Recommended Dilutions: ELISA: 1:15000-1:65000
Type: Primary
Antigen: IBTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human