Anti-GSK3B Rabbit Polyclonal Antibody
Catalog # ABNOPAB10055
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glycogen synthase kinase 3 beta
- Antigen symbol:GSK3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Zebrafish,Frog,Chicken,Human,Dog,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2932
- Antigen synonyms:EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
- Modification:phospho-Ser9
- Storage buffer:20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S9 of human GSK3B.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B.
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:3000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000
Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Dog, Frog, Human, Rat, Zebrafish