Anti-MDC1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB0366
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mediator of DNA Damage Checkpoint 1
- Antigen symbol:MDC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9656
- Antigen synonyms:6820401C03|AA413496|mKIAA0170|Nfbd1
- Amino acid number:871 to 888
- Storage buffer:PBS (0,2% gelatin, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 871-888 of human MDC1.
- Size:200 μl
- Pk:200 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MDC1.
Type: Primary
Antigen: MDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human